siRNA Details
| Gene Symbol: Suds3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122666.2 | NP_001116138.1 | 7, 8 | 800 | ||
| NM_178622.5 | NP_848737.3 | 7, 8 | 800 | |||
| GenBank mRNA | AF469109.1 | - | 7, 8 | 740 | ||
| AK010274.1 | - | 7, 8 | 704 | |||
| AK043652.1 | - | 7, 8 | 743 | |||
| AK045475.1 | - | 7, 8 | 754 | |||
| AK047910.1 | - | 1, 2 | 133 | |||
| AK138947.1 | - | 7, 8 | 751 | |||
| AK147512.1 | - | 1, 2 | 3808 | |||
| AK150633.1 | - | 7, 8 | 738 | |||
| BC036562.1 | - | 2, 3 | 173 | |||
| BC040088.1 | - | 5, 6 | 413 | |||
| BC055764.1 | - | 7, 8 | 753 | 6402735 | ||
| BC062176.1 | - | 7, 8 | 667 | 6514171 | ||

Suds3
Genomic Map