siRNA Details
| Gene Symbol: Tmco6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028036.3 | NP_082312.3 | 8, 9 | 1008 | ||
| GenBank mRNA | AK010487.1 | - | 8, 9 | 933 | ||
| AK046207.1 | - | 8, 9 | 1046 | |||
| BC005637.1 | - | 8, 9 | 1010 | 3156734 | ||

Tmco6
Genomic Map