siRNA Details
| Gene Symbol: Sars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023637.3 | NP_076126.2 | 11, 12 | 1096 | ||
| GenBank mRNA | AB029949.1 | - | 11, 12 | 1068 | ||
| AK010491.1 | - | 11, 12 | 1074 | |||
| AK087606.1 | - | 11, 12 | 1063 | |||
| AK140760.1 | - | 11, 12 | 1063 | |||
| AK178578.1 | - | 5, 6 | 371 | |||
| BC079664.1 | - | 11, 12 | 1073 | 30357834 | ||

Sars2
Genomic Map