siRNA Details
| Gene Symbol: Cyb5r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028057.2 | NP_082333.1 | 8 | 746 | ||
| GenBank mRNA | AK005159.1 | - | 8 | 747 | ||
| AK159663.1 | - | 8 | 716 | |||
| AK167178.1 | - | 8 | 740 | |||
| BC016266.1 | - | 8 | 1542 | 4924006 | ||
| BC024618.1 | - | 8 | 724 | 4235599 | ||
| JX420288.1 | - | 826 | ||||

Cyb5r1
Genomic Map