siRNA Details
| Gene Symbol: Strada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252448.1 | NP_001239377.1 | 12 | 1306 | ||
| NM_028126.3 | NP_082402.1 | 10 | 1195 | |||
| GenBank mRNA | AK011451.1 | - | 10 | 1141 | ||
| AK138358.1 | - | 12 | 1307 | |||
| AY341872.1 | - | 10 | 1154 | |||
| BC052063.1 | - | 1202 | ||||
| BC058517.1 | - | 10 | 1096 | 5717255 | ||

Strada
Genomic Map