siRNA Details
| Gene Symbol: 1700030K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089918.1 | - | 5 | 1284 | ||
| BC008558.1 | - | 5 | 1296 | 3592293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700030K09Rik| Gene Symbol: 1700030K09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK089918.1 | - | 5 | 1284 | ||
| BC008558.1 | - | 5 | 1296 | 3592293 | ||