siRNA Details
| Gene Symbol: Klhl40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028202.3 | NP_082478.1 | 1 | 1295 | ||
| GenBank mRNA | AK009491.1 | - | 1 | 1211 | ||
| AK078669.1 | - | 1 | 1209 | |||
| AK160799.1 | - | 1 | 1155 | |||
| BC125576.1 | - | 1 | 1295 | 40130091 | ||
| BC137889.1 | - | 1 | 1295 | 8860908 | ||

Klhl40
Genomic Map