siRNA Details
| Gene Symbol: Ripk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023663.6 | NP_076152.2 | 8 | 1541 | ||
| GenBank mRNA | AF302127.3 | - | 8 | 1544 | ||
| AK145203.1 | - | 8 | 1523 | |||
| BC057871.1 | - | 8 | 1504 | 5294197 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ripk4| Gene Symbol: Ripk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023663.6 | NP_076152.2 | 8 | 1541 | ||
| GenBank mRNA | AF302127.3 | - | 8 | 1544 | ||
| AK145203.1 | - | 8 | 1523 | |||
| BC057871.1 | - | 8 | 1504 | 5294197 | ||