siRNA Details
| Gene Symbol: Tspyl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030203.2 | NP_084479.1 | 1 | 307 | ||
| GenBank mRNA | AK020999.1 | - | 1 | 218 | ||
| AK030740.1 | - | 1 | 252 | |||
| AK034070.1 | - | 1 | 274 | |||
| AK034414.1 | - | 1 | 248 | |||
| AK038624.1 | - | 1178 | ||||
| AK138536.1 | - | 1 | 1087 | |||
| AK158197.1 | - | 1 | 275 | |||
| BC017540.1 | - | 1 | 256 | 4511620 | ||
| BC034656.1 | - | 1 | 255 | 5361000 | ||
| BY248195.1 | - | 1 | 308 | |||

Tspyl4
Genomic Map