siRNA Details
| Gene Symbol: Acbd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028250.3 | NP_082526.2 | 4 | 952 | ||
| GenBank mRNA | AK011781.1 | - | 4 | 945 | ||
| AK167959.1 | - | 5 | 892 | |||
| AK209754.1 | - | 4 | 267 | |||
| BC115580.1 | - | 4 | 544 | 40102241 | ||
| BC115581.1 | - | 4 | 544 | 40102242 | ||

Acbd6
Genomic Map