siRNA Details
Gene Symbol: Tmem161b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175187.5 | NP_780396.2 | 12 | 1425 | ||
GenBank mRNA | AK013305.1 | - | 11 | 1254 | ||
AK088393.1 | - | 12 | 1397 | |||
BC037611.1 | - | 10 | 1205 | |||
BC051663.1 | - | 12 | 1373 | 30020101 | ||
BC060067.1 | - | 12 | 1467 |
