siRNA Details
| Gene Symbol: Arhgap8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164627.1 | NP_001158099.1 | 10, 11 | 877 | ||
| NM_001164628.1 | NP_001158100.1 | 9, 10 | 869 | |||
| NM_001205334.1 | NP_001192263.1 | 10, 11 | 897 | |||
| NM_028455.4 | NP_082731.2 | 9, 10 | 889 | |||
| GenBank mRNA | AK014171.1 | - | 9, 10 | 840 | ||
| AK086816.1 | - | 9, 10 | 843 | |||
| AK088242.1 | - | 9, 10 | 845 | |||
| BC005563.1 | - | 8, 9 | 832 | 3965919 | ||
| BC010306.1 | - | 9, 10 | 859 | |||

Arhgap8
Genomic Map