siRNA Details
| Gene Symbol: Setd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080793.5 | NP_542983.3 | 5 | 885 | ||
| GenBank mRNA | AF448509.1 | - | 5 | 667 | ||
| AK048924.1 | - | 5 | 722 | |||
| AK129428.1 | - | 5 | 674 | |||
| AK147316.1 | - | 5 | 941 | |||
| AK147413.1 | - | 5 | 711 | |||
| AK147422.1 | - | 5 | 886 | |||
| AK147667.1 | - | 5 | 920 | |||
| AK170161.1 | - | 5 | 729 | |||
| BC050190.1 | - | 5 | 709 | 30007831 | ||

Setd7
Genomic Map