siRNA Details
Gene Symbol: Ccsap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028536.1 | NP_082812.1 | 4 | 1018 | ||
GenBank mRNA | AK006786.1 | - | 3 | 410 | ||
AK086180.1 | - | 3 | 724 | |||
BC024705.1 | - | 3 | 675 | 3668303 | ||
BC036300.1 | - | 4 | 1018 | 4501054 |
