siRNA Details
| Gene Symbol: Glipr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028608.3 | NP_082884.1 | 2 | 617 | ||
| GenBank mRNA | AK010768.1 | - | 2 | 549 | ||
| AK137557.1 | - | 2 | 564 | |||
| AK170051.1 | - | 2 | 564 | |||
| BC025083.1 | - | 2 | 541 | 3711757 | ||
| BC098231.1 | - | 2 | 591 | 3325823 | ||

Glipr1
Genomic Map