siRNA Details
Gene Symbol: Mvb12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028617.2 | NP_082893.1 | 4 | 410 | ||
GenBank mRNA | AK003644.1 | - | 4 | 390 | ||
BC020119.1 | - | 4 | 410 | 3982577 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mvb12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028617.2 | NP_082893.1 | 4 | 410 | ||
GenBank mRNA | AK003644.1 | - | 4 | 390 | ||
BC020119.1 | - | 4 | 410 | 3982577 |