siRNA Details
Gene Symbol: Mvb12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028617.2 | NP_082893.1 | 3 | 260 | ||
GenBank mRNA | AK003644.1 | - | 3 | 240 | ||
BC020119.1 | - | 3 | 260 | 3982577 | ||
BC027419.1 | - | 3 | 236 | 4163554 | ||
BC087734.1 | - | 3 | 257 | 5101734 | ||
BC096439.1 | - | 2 | 417 |
