siRNA Details
| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028634.3 | NP_082910.1 | 4, 5 | 447 | ||
| GenBank mRNA | AF331040.1 | - | 4, 5 | 447 | ||
| AK003719.1 | - | 4, 5 | 423 | |||
| BC005733.1 | - | 3, 4 | 400 | 3596486 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cby1| Gene Symbol: Cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028634.3 | NP_082910.1 | 4, 5 | 447 | ||
| GenBank mRNA | AF331040.1 | - | 4, 5 | 447 | ||
| AK003719.1 | - | 4, 5 | 423 | |||
| BC005733.1 | - | 3, 4 | 400 | 3596486 | ||