siRNA Details
| Gene Symbol: Whrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008791.2 | NP_001008791.1 | 7 | 2307 | ||
| NM_001008792.2 | NP_001008792.1 | 7 | 2319 | |||
| NM_001008793.2 | NP_001008793.1 | 7 | 2307 | |||
| NM_028640.2 | NP_082916.1 | 7 | 2307 | |||
| GenBank mRNA | AK021254.1 | - | 1 | 73 | ||
| AK122523.1 | - | 7 | 1438 | |||
| AK157955.1 | - | 7 | 2308 | |||
| AY739114.1 | - | 7 | 2267 | |||
| AY739115.1 | - | 7 | 2267 | |||
| AY739116.1 | - | 7 | 2279 | |||
| AY739117.1 | - | 7 | 2267 | |||
| AY739118.1 | - | 3 | 883 | |||
| AY739119.1 | - | 3 | 883 | |||
| AY739120.1 | - | 2 | 670 | |||
| AY739121.1 | - | 2 | 788 | |||
| AY739122.1 | - | 2 | 788 | |||
| BC150757.1 | - | 7 | 1664 | 9087668 | ||
| HQ148552.1 | - | 7 | 1549 | |||

Whrn
Genomic Map