siRNA Details
| Gene Symbol: Fam161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028672.2 | NP_082948.2 | 3 | 758 | ||
| GenBank mRNA | AK133447.1 | - | 3 | 774 | ||
| BC024460.1 | - | 4 | 691 | 4975427 | ||
| BC026495.1 | - | 3 | 758 | 4505506 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam161a| Gene Symbol: Fam161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028672.2 | NP_082948.2 | 3 | 758 | ||
| GenBank mRNA | AK133447.1 | - | 3 | 774 | ||
| BC024460.1 | - | 4 | 691 | 4975427 | ||
| BC026495.1 | - | 3 | 758 | 4505506 | ||