siRNA Details
| Gene Symbol: Fam161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028672.2 | NP_082948.2 | 4 | 1699 | ||
| GenBank mRNA | BC024460.1 | - | 5 | 1632 | 4975427 | |
| BC026495.1 | - | 4 | 1699 | 4505506 | ||
| BQ032025.1 | - | 1 | 251 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam161a| Gene Symbol: Fam161a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028672.2 | NP_082948.2 | 4 | 1699 | ||
| GenBank mRNA | BC024460.1 | - | 5 | 1632 | 4975427 | |
| BC026495.1 | - | 4 | 1699 | 4505506 | ||
| BQ032025.1 | - | 1 | 251 | |||