siRNA Details
| Gene Symbol: Ift57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028680.3 | NP_082956.2 | 4 | 589 | ||
| GenBank mRNA | AF274590.1 | - | 4 | 607 | ||
| AK047217.1 | - | 4 | 589 | |||
| BC138143.1 | - | 4 | 563 | 8861162 | ||
| BC138144.1 | - | 4 | 563 | 8861163 | ||
| DQ266091.1 | - | 4 | 630 | |||

Ift57
Genomic Map