siRNA Details
| Gene Symbol: Phf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028716.4 | NP_082992.1 | 2 | 189 | ||
| GenBank mRNA | AK014030.1 | - | 2 | 238 | ||
| AK014380.1 | - | 2 | 190 | |||
| BC027776.1 | - | 2 | 184 | 5352251 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Phf19| Gene Symbol: Phf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028716.4 | NP_082992.1 | 2 | 189 | ||
| GenBank mRNA | AK014030.1 | - | 2 | 238 | ||
| AK014380.1 | - | 2 | 190 | |||
| BC027776.1 | - | 2 | 184 | 5352251 | ||