siRNA Details
| Gene Symbol: Ehhadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023737.3 | NP_076226.2 | 7 | 1476 | ||
| GenBank mRNA | AJ011864.1 | - | 1 | 120 | ||
| AK004867.1 | - | 7 | 1447 | |||
| BC016899.1 | - | 7 | 1450 | 4241677 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ehhadh| Gene Symbol: Ehhadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023737.3 | NP_076226.2 | 7 | 1476 | ||
| GenBank mRNA | AJ011864.1 | - | 1 | 120 | ||
| AK004867.1 | - | 7 | 1447 | |||
| BC016899.1 | - | 7 | 1450 | 4241677 | ||