siRNA Details
| Gene Symbol: Iqce | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028833.3 | NP_083109.2 | 20, 21 | 2075 | ||
| GenBank mRNA | AK006472.2 | - | 19, 20 | 1816 | ||
| BC059223.1 | - | 20, 21 | 2075 | 6417270 | ||
| BC065128.1 | - | 16, 17 | 4640 | |||

Iqce
Genomic Map