siRNA Details
| Gene Symbol: Usp20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028846.5 | NP_083122.1 | 22 | 2563 | ||
| GenBank mRNA | AF449715.1 | - | 22 | 2514 | ||
| AK006800.1 | - | 5 | 497 | |||
| AK029890.1 | - | 2459 | ||||
| AK047242.1 | - | 22 | 2482 | |||
| AK054279.1 | - | 22 | 2506 | |||
| AK087412.1 | - | 22 | 2475 | |||
| AK163663.1 | - | 22 | 2473 | |||
| AK173083.1 | - | 22 | 2465 | |||
| AK215664.1 | - | 1 | 13 | |||
| BC019735.1 | - | 8 | 763 | |||
| BC037199.1 | - | 16 | 1947 | |||
| BC079674.1 | - | 2452 | 30531994 | |||

Usp20
Genomic Map