siRNA Details
Gene Symbol: Ttll9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083618.1 | NP_001077087.1 | 3 | 155 | ||
NM_029064.2 | NP_083340.2 | 3 | 155 | |||
GenBank mRNA | AK006027.1 | - | 3 | 155 | ||
AK015388.1 | - | 3 | 135 | |||
AK015740.1 | - | 3 | 135 | |||
AM690753.1 | - | 2 | 99 | |||
BC130024.1 | - | 3 | 139 | |||
BC148285.2 | - | 2 | 95 |
