siRNA Details
Gene Symbol: Rab3il1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144538.2 | NP_653121.1 | 4, 5 | 568 | ||
GenBank mRNA | AK132764.1 | - | 7, 8 | 1275 | ||
AK161298.1 | - | 7, 8 | 1275 | |||
BC020147.1 | - | 4, 5 | 533 | 3708973 |
