siRNA Details
| Gene Symbol: Atp6v1e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029121.3 | NP_083397.3 | 2 | 407 | ||
| GenBank mRNA | AB074757.1 | - | 2 | 394 | ||
| AK015654.1 | - | 2 | 394 | |||
| BC049547.1 | - | 2 | 395 | 6770428 | ||
| BC061059.1 | - | 2 | 407 | 6744043 | ||

Atp6v1e2
Genomic Map