siRNA Details
| Gene Symbol: Pmis2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB690425.1 | - | 1 | 93 | ||
| AK015595.1 | - | 1 | 151 | |||
| BC100424.1 | - | 1 | 157 | 30920315 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_183108.1 | - | 151 | Chr. - - [-] on Build | ||

Pmis2
Genomic Map