siRNA Details
| Gene Symbol: Mettl5os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016085.1 | - | 1 | 205 | ||
| AK050748.1 | - | 1 | 274 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl5os| Gene Symbol: Mettl5os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016085.1 | - | 1 | 205 | ||
| AK050748.1 | - | 1 | 274 | |||