siRNA Details
| Gene Symbol: 1700001C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029285.1 | NP_083561.1 | 3 | 430 | ||
| GenBank mRNA | AK005574.1 | - | 3 | 430 | ||
| BC060996.1 | - | 3 | 433 | 6743526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700001C02Rik| Gene Symbol: 1700001C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029285.1 | NP_083561.1 | 3 | 430 | ||
| GenBank mRNA | AK005574.1 | - | 3 | 430 | ||
| BC060996.1 | - | 3 | 433 | 6743526 | ||