siRNA Details
| Gene Symbol: 1700019G17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145895.1 | NP_001139367.1 | 3 | 799 | ||
| NM_029331.3 | NP_083607.3 | 4 | 822 | |||
| GenBank mRNA | AK006120.1 | - | 3 | 800 | ||
| BC029200.1 | - | 768 | 3501234 | |||

1700019G17Rik
Genomic Map