siRNA Details
Gene Symbol: Ssmem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027073.1 | NP_081349.1 | 3 | 938 | ||
NM_029373.4 | NP_083649.1 | 3 | 752 | |||
GenBank mRNA | AK006038.1 | - | 3 | 939 | ||
AK006332.1 | - | 3 | 738 | |||
BC049755.1 | - | 3 | 887 | 6773255 |
