siRNA Details
| Gene Symbol: Rasl10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145216.3 | NP_660251.1 | 2 | 420 | ||
| GenBank mRNA | AF498251.1 | - | 2 | 242 | ||
| AK044011.1 | - | 2 | 421 | |||
| BC107200.1 | - | 3 | 303 | |||
| BC107201.1 | - | 3 | 301 | 40051192 | ||

Rasl10a
Genomic Map