siRNA Details
| Gene Symbol: Mff | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029409.2 | NP_083685.2 | 3 | 328 | ||
| GenBank mRNA | AK017226.1 | - | 3 | 328 | ||
| AK083056.1 | - | 3 | 313 | |||
| AK139649.1 | - | 3 | 307 | |||
| AK218934.1 | - | 3 | 185 | |||
| BC016597.1 | - | 1 | 39 | 3489844 | ||
| BC059229.1 | - | 3 | 328 | 6413193 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001271284, NM_001276401, NM_001039015, XM_006245204, XM_008767232, and others.

Mff
Genomic Map