siRNA Details
| Gene Symbol: Hormad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029458.1 | NP_083734.1 | 10, 11 | 1056 | ||
| GenBank mRNA | AK015939.1 | - | 11, 12 | 1058 | ||
| BC120781.1 | - | 10, 11 | 1056 | 40129704 | ||
| BC120783.1 | - | 10, 11 | 1056 | 40129706 | ||
| BC144842.1 | - | 10, 11 | 1056 | 9053390 | ||

Hormad2
Genomic Map