siRNA Details
| Gene Symbol: Zfp773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029584.1 | NP_083860.1 | 4 | 1133 | ||
| GenBank mRNA | AK013056.1 | - | 4 | 1101 | ||
| AK031040.1 | - | 4 | 1133 | |||
| BC117497.1 | - | 4 | 1111 | 40106595 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp773| Gene Symbol: Zfp773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029584.1 | NP_083860.1 | 4 | 1133 | ||
| GenBank mRNA | AK013056.1 | - | 4 | 1101 | ||
| AK031040.1 | - | 4 | 1133 | |||
| BC117497.1 | - | 4 | 1111 | 40106595 | ||