siRNA Details
| Gene Symbol: Zfp735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126489.2 | NP_001119961.1 | 4 | 1569 | ||
| GenBank mRNA | AK145370.1 | - | 4 | 1570 | ||
| BC137974.1 | - | 4 | 1499 | 8860993 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp735| Gene Symbol: Zfp735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126489.2 | NP_001119961.1 | 4 | 1569 | ||
| GenBank mRNA | AK145370.1 | - | 4 | 1570 | ||
| BC137974.1 | - | 4 | 1499 | 8860993 | ||