siRNA Details
Gene Symbol: Zfp735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126489.2 | NP_001119961.1 | 4 | 1398 | ||
GenBank mRNA | AK145370.1 | - | 4 | 1399 | ||
BC137974.1 | - | 4 | 1328 | 8860993 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126489.2 | NP_001119961.1 | 4 | 1398 | ||
GenBank mRNA | AK145370.1 | - | 4 | 1399 | ||
BC137974.1 | - | 4 | 1328 | 8860993 |