siRNA Details
| Gene Symbol: Rftn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181397.2 | NP_852062.1 | 7 | 1349 | ||
| GenBank mRNA | AK082517.1 | - | 962 | |||
| AK087803.1 | - | 1123 | ||||
| AK088466.1 | - | 5 | 886 | |||
| AK152130.1 | - | 7 | 1271 | |||
| AK153288.1 | - | 7 | 1271 | |||
| AK155870.1 | - | 7 | 1313 | |||
| AK170913.1 | - | 7 | 1312 | |||
| AK172884.1 | - | 7 | 1277 | |||
| BC043324.1 | - | 3 | 406 | |||
| BC052074.1 | - | 7 | 1349 | 6401192 | ||

Rftn1
Genomic Map