siRNA Details
| Gene Symbol: Fbxo31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133765.4 | NP_598526.3 | 4 | 572 | ||
| GenBank mRNA | AB041586.1 | - | 4 | 421 | ||
| AK009859.1 | - | 1 | 13 | |||
| AK140412.1 | - | 4 | 573 | |||
| AK163641.1 | - | 4 | 541 | |||
| AK197038.1 | - | 1 | 21 | |||
| BC026929.1 | - | 4 | 557 | 4194857 | ||

Fbxo31
Genomic Map