siRNA Details
Gene Symbol: Spatc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029661.1 | NP_083937.1 | 4 | 1030 | ||
GenBank mRNA | AK006232.1 | - | 4 | 935 | ||
AK006410.1 | - | 4 | 1030 | |||
BC049686.1 | - | 4 | 1022 | |||
BC147585.1 | - | 4 | 1010 | 9056575 | ||
BC147614.1 | - | 4 | 1010 | 9056604 |
