siRNA Details
| Gene Symbol: Mib2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256107.1 | NP_001243036.1 | 3 | 1048 | ||
| NM_001256108.2 | NP_001243037.2 | 4 | 964 | |||
| NM_145124.3 | NP_660106.2 | 3 | 1048 | |||
| GenBank mRNA | AB063290.1 | - | 1 | 51 | ||
| AB072336.1 | - | 3 | 610 | |||
| AK147940.1 | - | 4 | 908 | |||
| AK159611.1 | - | 3 | 766 | |||
| AY974090.1 | - | 1 | 189 | |||
| BC058086.1 | - | 2 | 307 | |||
| BC141133.1 | - | 2 | 362 | 9055745 | ||
| BC145441.1 | - | 2 | 362 | |||

Mib2
Genomic Map