siRNA Details
| Gene Symbol: Wbscr28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029681.3 | NP_083957.1 | 2 | 149 | ||
| NM_194065.3 | NP_919041.1 | 2 | 149 | |||
| GenBank mRNA | AK007170.1 | - | 2 | 150 | ||
| AY369079.1 | - | 2 | 150 | |||
| AY369080.1 | - | 2 | 150 | |||
| BC114542.1 | - | 2 | 150 | 40050280 | ||

Wbscr28
Genomic Map