siRNA Details
| Gene Symbol: 2410124H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029740.1 | NP_084016.1 | 2 | 372 | ||
| GenBank mRNA | AK010774.1 | - | 2 | 373 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2410124H12Rik| Gene Symbol: 2410124H12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029740.1 | NP_084016.1 | 2 | 372 | ||
| GenBank mRNA | AK010774.1 | - | 2 | 373 | ||