siRNA Details
| Gene Symbol: Dok5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163686.1 | NP_001157158.1 | 2 | 401 | ||
| NM_029761.4 | NP_084037.3 | 2 | 401 | |||
| GenBank mRNA | AF418208.1 | - | 2 | 263 | ||
| AK012430.1 | - | 2 | 77 | |||
| AK044148.1 | - | 2 | 269 | |||
| BC066822.1 | - | 2 | 401 | 30021902 | ||
| BY248468.1 | - | 2 | 317 | |||

Dok5
Genomic Map