siRNA Details
Gene Symbol: Kdm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029842.5 | NP_084118.1 | 8 | 1352 | ||
GenBank mRNA | AK014006.1 | - | 8 | 1296 | ||
BC024807.1 | - | 8 | 1280 | 5355270 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kdm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029842.5 | NP_084118.1 | 8 | 1352 | ||
GenBank mRNA | AK014006.1 | - | 8 | 1296 | ||
BC024807.1 | - | 8 | 1280 | 5355270 |