siRNA Details
| Gene Symbol: 1700109H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029843.2 | NP_084119.1 | 3 | 276 | ||
| GenBank mRNA | AK007150.1 | - | 3 | 319 | ||
| BC048606.1 | - | 3 | 426 | |||
| BC099581.1 | - | 3 | 276 | 30920581 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700109H08Rik| Gene Symbol: 1700109H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029843.2 | NP_084119.1 | 3 | 276 | ||
| GenBank mRNA | AK007150.1 | - | 3 | 319 | ||
| BC048606.1 | - | 3 | 426 | |||
| BC099581.1 | - | 3 | 276 | 30920581 | ||