siRNA Details
| Gene Symbol: Cutal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021330.1 | - | 6 | 505 | ||
| AK033810.1 | - | 6 | 583 | |||
| AK085616.1 | - | 6 | 544 | |||
| BC021771.1 | - | 5 | 410 | 4194825 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cutal| Gene Symbol: Cutal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021330.1 | - | 6 | 505 | ||
| AK033810.1 | - | 6 | 583 | |||
| AK085616.1 | - | 6 | 544 | |||
| BC021771.1 | - | 5 | 410 | 4194825 | ||